Titanium 99,2 Metals basis, foil, Thickness: 1.0 mm (0.040 in)

Supplier: Thermo Fisher Scientific
Warning

44484.GHEA 91.9 EUR
44484.GH 44484.GW
Titanium 99,2 Metals basis, foil, Thickness: 1.0 mm (0.040 in)
Titanium

Purity: 99.2% CAS No.: 7440-32-6 Physical Form: ≈45g/100x100mm Odour: Odourless MDL No.: MFCD00011264

Formula: Ti
MW: 47.88 g/mol
Boiling Pt: 3262
Melting Pt: 1660
Density: 4.51 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011264
CAS Number: 7440-32-6
EINECS: 231-142-3
Merck Index: 12,09610

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR