Indium tin oxide ≥99.5%, NanoTek®

Supplier: Thermo Fisher Scientific

Synonyms: ITO

44927.14 44927.22
44927.14EA 267 EUR
44927.14 44927.22
Indium tin oxide ≥99.5%, NanoTek®
Indium tin oxide

Applications: Indium tin oxide is one of the most widely used transparent conducting oxides due to its important properties like electrical conductivity and optical transparency, as well as the easy formation of thin film on deposition. It is also used in touch panels, electrochromic, electroluminescent and LCD displays, plasma displays, field emission displays, heat reflective coatings, energy efficient windows, gas sensors and photovoltaics.


Applications: NanoTek™ indium tin oxide can be used to impart conductivity to transparent coatings such as those on the surface of computer display tubes or on the films used to produce conductive display screens such as for interactive touch screens

Formula: In₂O₃·(SnO₂)x
Density: 7.16
Storage Temperature: Ambient
MDL Number: MFCD00171662
CAS Number: 50926-11-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR