PTFE, sheets, Thickness: 3.18 mm (0.125 in)

Supplier: Thermo Fisher Scientific

Synonyms: Polytetrafluoroethylene, Teflon

45172.HB
45172.HBEA 192 EUR
45172.HB
PTFE, sheets, Thickness: 3.18 mm (0.125 in)
PTFE

MDL No.: MFCD00084455

Formula: (CF₂CF₂)n
Storage Temperature: Ambient
MDL Number: MFCD00084455
CAS Number: 9002-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR