Nickel standard solution, 10 mg/L Ni in 5% nitric acid (from Ni), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific

45261.AEEA 89.8 EUR
45261.AE
Nickel standard solution, 10 mg/L Ni in 5% nitric acid (from Ni), standard for ICP, Specpure®
Standards Cationic Standards Nickel Standards

Molecular Formula: Ni in 2% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011137


Certifikace: Traceable to NIST

Order Now

SPECIFICATIONS

Environmentally Preferable
MDL No. MFCD00011137

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR