TR Ovens with Safety Technology for Charges Containing Solvents acc. to EN 1539
Supplier: NABERTHERM
TRS065ON TRS125ON TRS245ON TRS455ON
455-2131EA 9980 EUR
455-2131 455-2132 455-2133 455-2134
TR Ovens with Safety Technology for Charges Containing Solvents acc. to EN 1539
Ovens Standard Ovens
Ovens that are designed for drying and evaporation of solvent-containing materials. Furnace based technology utilised for ovens, units have a 1.4301 (DIN) stainless steel chamber, which is rust-resistant and easy to clean, they feature horizontal forced convection.
  • Required continuous air exchange in the oven which is achieved via forced convection fan
  • Temperature limit controller with adjustable cut-out temperature for thermal protection class 2 in accordance to EN 60519-2 for protection of the oven and load
  • Differential pressure monitoring of the exhaust