VWR®, Calcium standard solution, 10000 mg/L Ca in dil. nitric acid (from CaCO₃) ARISTAR® standard for ICP
Supplier: VWR Chemicals
455172TEA 203 EUR
455172T 455174V
VWR®, Calcium standard solution, 10000 mg/L Ca in dil. nitric acid (from CaCO₃) ARISTAR® standard for ICP
Standards Cationic Standards Calcium Standards
CaCO₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.