VWR® ARISTAR®, Lutetium Standard Solution, 1000 mg/L Lu in dil. Nitric Acid (from Lu₂O₃), Standard for ICP
Supplier: VWR Chemicals
455602UEA 537 EUR
455602U 455604W
VWR® ARISTAR®, Lutetium Standard Solution, 1000 mg/L Lu in dil. Nitric Acid (from Lu₂O₃), Standard for ICP
Standards Cationic Standards Lutetium Standards
Lu₂O₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.