VWR® ARISTAR®, Multi-Element Calibration Standard 4 in Water with a Trace of Hydrofluoric Acid for ICP-MS
Supplier: VWR Chemicals
456522CEA 450 EUR
456522C
VWR® ARISTAR®, Multi-Element Calibration Standard 4 in Water with a Trace of Hydrofluoric Acid for ICP-MS
Standards Cationic Standards Multi-Element Standards
(12 Elements in H₂O with trace of HF ).
12 Elements 10 mg/L : B, Ge, Mo, Nb, P, Re, S, Si, Ta, Ti, W, Zr
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.