Thermoshaker for deep well plates, TS-DW
Supplier: GRANT INSTRUMENTS
TS-DW
460-0324EA 1691 EUR
460-0324
Thermoshaker for deep well plates, TS-DW
Block Heaters Shaking Block Heaters
The TS-DW has been designed for shaking and heating deep well plates. A multisystem principle used in the design of the thermoshaker allows it to operate as three independent devices: Incubator, plate shaker and thermoshaker. Ideal for use in life science applications, molecular biology, cell biology lab, cell lysis, DNA isolation and purification, sample preparation for PCR, pellet re-suspension, or any other method where many samples need mixing in deep well plates.
  • Profiled platform for perfect plate fit and maximum heat transfer
  • Rapid heat-up speed
More Product Information
Order Now
SPECIFICATIONS
Model TS-DW
Environmentally Preferable
Orbit 2 mm
Temperature control range Ambient +5...100 °C
Heating speed Using Eppendorf block:
from 25...100 °С: 4 °С/min,
from 25...37 ºС: 8 min
Temperature accuracy ±0,5 °C
W×D×H 245×255×165 mm
Weight 5,1 kg
Speed range 250 - 1400 min⁻¹