Baths and lids, stainless steel, ST series
Supplier: GRANT INSTRUMENTS
ST26 ST12 ST18 ST38 ST5 G-BTL G-BTS LST12 LST26 LST38
461-0674EA 840 EUR
461-0674 461-0672 461-0673 461-0675 461-0676 GRANG-BTL GRANG-BTS GRANLST12 GRANLST26 GRANLST38
Baths and lids, stainless steel, ST series
Baths Water Baths, Unstirred
Premium baths which are constructed from corrosion resistant stainless steel for durability in demanding environments. Available in five sizes: 5, 12, 18, 26 and 38 litres to suit a wide variety of applications.
  • All baths are supplied with mounting bridge plate. Models of 12 litres and above have a mounting plate which allows the Optima™ thermostat to be orientated on the tank in two directions - ideal for optimising bench space
  • Baths of 12 litres and above feature a drain tap for easy emptying and can accommodate Grant C1G and C2G immersion coolers for