Heating circulators, CORIO™ CD-BC series
Supplier: JULABO GmbH
9012504 9012506 9012512 9012526
461-0916EA 1989 EUR
461-0916 461-0917 461-0918 461-0919
Heating circulators, CORIO™ CD-BC series
Circulating Thermostats Open Circulator Systems Baths with Integral Circulators
The powerful and robust CORIO™ laboratory circulators provide the exact temperature, absolute precision, and a wide working temperature range. The CORIO™ CD model is an advanced unit with professional technology for demanding applications and can be used for temperature control of internal or external applications. The jet nozzle allows continuous adjustment of the pump stream in the system. The insulated stainless steel baths are durable and have an integrated drain tap. These heating circulators are optimised for external temperature applications in combination with jacketed reactors,More Product Information