VWR®, Refrigerated Circulators
Supplier: VWR Collection
462-0229EA 7840 EUR
462-0229 462-0238 462-0237 462-0236 462-0235 462-0234 462-0233 462-0232 462-0230 462-0228 462-0227 462-0226 462-0224 462-0239 462-0231
VWR®, Refrigerated Circulators
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Stainless steel circulating baths with choice of Advanced Programmable or Advanced Digital temperature controller. Both are easy to navigate with large, intuitive displays and multiple communication options, including USB-A and B, RS232/485, Ethernet and external temperature probe. The reservoir drain can be accessed by removing the front panel. All models feature user-adjustable, high temperature safety cut-off points, as well as over-temperature protection. The surface is cool to the touch, even when operating at high temperatures.
  • Working temperatures from −40 to +200 °C with