VWR®, Circulating Chillers
Supplier: VWR Collection
462-7033EA 0 EUR
462-7033 462-7035 462-7040 462-7041
VWR®, Circulating Chillers
Circulating Thermostats Closed Circulator Systems Cooling Circulators
These circulating chillers are an environmentally friendly, water-conserving and reliable means of controlling the temperature of liquids in open tanks or closed loop systems. They provide high cooling performance at low cost. With a temperature range from −10 to +40 °C, they are ideally suited for many process and routine applications.
  • Exceptional cooling performance and wide temperature range from −10 to +40 °C
  • Extra large digital display for easy reading of the set temperature, the flow rate and/or the pressure even from a distance
  • Optical and acoustic
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Water Efficient