Secnidazole 98%

Supplier: Thermo Fisher Scientific

Synonyms: 1-(2-Methyl-5-nitroimidazol-1-yl)-2-propanol, 1-(2-Methyl-5-nitro-1H-imidazol-1-yl)propan-2-ol

469030010 469030050
469030010.EA 22.8 EUR
469030010. 469030050.
Secnidazole 98%
Secnidazole
Formula: C₇H₁₁N₃O₃
MW: 185.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00864656
CAS Number: 3366-95-8
EINECS: 222-134-0
Merck Index: 13,08494
New Product

Order Now

Specification Test Results

Appearance white to light yellow powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 70°C to 76°C
Water ≤6 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR