Pharmaceutical refrigerator, MED-600-S
Supplier: KIRSCH PHILIPP
10115
471-2539EA 0 EUR
471-2539
Pharmaceutical refrigerator, MED-600-S
Refrigerators
Thanks to its long drawers this pharmaceutical fridge is ideal for complementing pharmacies’ furniture. The drawers have a transparent bottom so users can see at a glimpse what is inside.
  • 12 height adjustable drawers, individual drawer division with adjustable length and cross dividers
  • Recirculating air-cooling
  • Optimised ventilation plates to minimise interior temperature differences
  • Automatic defrosting; melt water evaporation
  • Easy-to-read LED temperature display; easy accessible control panel with key switch
  • Micro-processor controlled temperature and