Recirculating coolers, FC series
Supplier: JULABO GmbH
9600063 9600123 9600126 9600160 9600060 9600120 9600163 9600166 9601256 9601060 9601063
473-3221EA 7772 EUR
473-3221 473-3231 473-3232 473-3240 473-3220 473-3230 473-3241 473-3242 473-0024 473-0038 473-0039
Recirculating coolers, FC series
Circulating Thermostats Closed Circulator Systems Cooling Circulators
Environmentally friendly recirculating coolers that save tap water, units are rapid and powerful, and offer short cool-down times and high efficiency. Due to the compact design, the coolers fit either on or under laboratory benchtops. FC units feature two separate multi-displays (LED) for actual and set point values.
  • Built-in heating element to allow cooling and heating applications with FC models
  • FC coolers have an adjustable ratio for inlet/outlet temperature
  • Connection for external PT100 sensor for external measurement and control (T-type models)
  • RS232