Water cooler
Supplier: SCOTSMAN
SCW14-B
474-0035EA 0 EUR
474-0035
Water cooler
Water Dispensers
The SWC14B water cooler has two independent taps, one controlled by a foot pedal the other is hand activated, both have automatic stream regulation. Unit is manufactured from stainless steel. Cooler can be free standing or wall tight to avoid unsightly connections.
  • Refrigerated drinking water fountain
  • Water thermostat positioned behind the front panel for ease of reach
  • Delivers 50 liters per hour (13.2 gph) of 8-12 °C (46-53 °F) water (at 32°C-90°F ambient and 21°C-70°F inlet water temperatures)
  • Easily removable front panel for routine maintenance and