Ice and water automatic hands-free dispenser
Supplier: SCOTSMAN
TC180
474-0036EA 0 EUR
474-0036
Ice and water automatic hands-free dispenser
Ice Makers
The TC180 ice cube and water dispenser is a fully automatic air cooled dispensing station that dispenses ice cubelets or water or both simultaneously in either measured amounts or continuously for filling vessels. The unit is manufactured from 304 stainless steel. Cubes are dispensed using a touch free dispensing system and the ice storage compartment is sealed ensuring that the ice cannot be contaminated. The ice cube/water dispenser requires a 19 mm ‘washing machine' type tapped mains water supply, a correctly rated power supply and a 38 mm vented drain (ideally no higher than the base ofMore Product Information