Ice flakers, self contained, EF series
Supplier: SCOTSMAN
EF103 EF124 EF156 EF206
474-0129EA 0 EUR
474-0129 474-0130 474-0131 474-0132
Ice flakers, self contained, EF series
Ice Makers
The EF self contained ice flakers are air-cooled and manufactured from 304 stainless steel.
  • Integral ice flake storage bin
  • Constructed of high grade stainless steel
  • Ideal for medical and laboratory use
All the models are automatic and produce small irregular flakes of ice with an even melting rate. The flakes are easily moulded to fit into any shape. The ice flakers require a 19 mm 'washing machine' type tapped mains water supply, a correctly rated power supply and a 38 mm vented drain (ideally no higher than the base of the machine), within one metreMore Product Information