Dry ice machines, SnowPack
Supplier: BURKLE
9805-0030 9805-0200
474-3110EA 269.7 EUR
474-3110 474-0057
Dry ice machines, SnowPack
Dry Ice Makers
SnowPack enables the user to obtain a −79 °C dry ice tablet within minutes, without complicated technology and without electricity. Can be installed on any commercial CO₂-bottle.
  • Ideal coolant for transporting
  • Dry ice is odourless and easy to handle