VWR® Cryo 1 °C Freezing Container
Supplier: VWR Collection
479-0640EA 99.9 EUR
479-0640
VWR® Cryo 1 °C Freezing Container
Boxes Cooling/Freezing Boxes and Bags
Container PC, closure and vial holder HDPE, foam insert.
  • Floating rigid insert prevents vials from contamination
  • Withstands repeated use; labelled with step-by-step instructions
  • Holds up to 18 cryogenic vials (1; 1,2; 1,5 and 2 ml)
Non mechanical device that uses isopropyl alcohol and a mechanical freezer to provide cooling rate of –1 °C per minute. Ideal for successful cell cryopreservation and recovery.