Caps for vials, Cryobank and Bank-It™, Nunc™
Supplier: Thermo Fisher Scientific
374018 374019 374021 374017
479-1074EA 185 EUR
479-1074 479-1075 479-1076 479-1073
Caps for vials, Cryobank and Bank-It™, Nunc™
Closures Cryogenic Vial Closures
Ensure contamination-free cap replacement with Thermo Scientific™ Nunc™ Cryobank and Bank-it™ Caps, which are supplied racked for manual or automated replacement.
  • Available in different colours for simplified sample tracking
  • Racked in 96 microplate pattern for easy contamination free pick
Limited Time Sale
Select items below are currently on sale