VWR® Sliding Tray Racks for Cryoboxes, Height 50 mm
Supplier: VWR Collection
479-2251EA 163 EUR
479-2251 479-2252 479-2253 479-2254 479-2255 479-2256 479-2257 479-2437
VWR® Sliding Tray Racks for Cryoboxes, Height 50 mm
Racks Cryogenic Box Racks
Freezer racks as storage and inventory systems for upright ULT freezers makes it easier organise your samples.
  • Different height and depth dimensions are available and can be used in all freezer brands
  • Racks for standard 2" cryoboxes (133×133×51 mm)
  • Made from corrosion resistant Stainless steel with smoothed edges
These racks ensure an ergonomic operating process as the trays are placed in the rack. With a pull-out design it makes it easy to get samples without taking the whole rack out from the freezer. The tray has an easy access handle.