VWR®, Test Sieves, 300×60 mm, with Square Holes
Supplier: VWR Collection
510-2789EA 281 EUR
510-2789 510-2790 510-2791 510-2792 510-2793 510-2794 510-2795 510-2796 510-2797 510-2798 510-2799 510-2801 510-2802 510-2803 510-2800 510-2804 510-2805 510-2806 510-2807 510-2808 510-2809 510-2810 510-2811 510-2812 510-2813 510-2814 510-2815 510-2816 510-2817 510-2818 510-2819 510-2820 510-2821 510-2822 510-2823 510-2824 510-2825 510-2826 510-2827 510-2828 510-2829
VWR®, Test Sieves, 300×60 mm, with Square Holes
Sieves Test Sieves
Frame: Stainless steel; perforated plates: Galvanised steel, ISO 3310-2, ASTM E323, BS 410, AFNOR.
Optional: Inspection certificate of calibration 3.1 according to DIN EN 10204 with measuring results of a specific test.
Sieves and accessories of other dimensions are available on request.
Please go to website for a full overview.
All sieves are supplied with serial number and a certificate of compliance with the order 2.1 according to DIN EN 10204. Further certificates and calibration services are available. Contact your local Avantor sales office for further details.