Kjeldahl weighing paper, grade B-2, Whatman™
Supplier: Whatman products (Cytiva)
10347671 10347673 10347670 10347672
512-0602EA 40.9 EUR
512-0602 512-0604 512-0601 512-0603
Kjeldahl weighing paper, grade B-2, Whatman™
Weighing Papers
This weighing paper dissolves residue free in acid solution, which is critical in applications such as Kjeldahl analysis of nitrogen in soil.
  • Transparent and smooth
  • Simplifies sample transfer
  • Quantitative transfer from paper