Micro Dialysis System, Tube-O-Dialyzer™
Supplier: G-Biosciences
786-617 786-612 786-610 786-611 786-613 786-614 786-615 786-616 786-618 786-619 786-620 786-621 786-622 786-623 786-624 786-613T 786-610T 786-616T 786-619T 786-617T 786-615T 786-614T 786-612T 786-611T 786-618T
514-0160EA 409 EUR
514-0160 514-0155 514-0153 514-0154 514-0156 514-0157 514-0158 514-0159 514-0161 514-0162 514-0163 514-0164 514-0165 514-0166 514-0167 786-613T 786-610T 786-616T 786-619T 786-617T 786-615T 786-614T 786-612T 786-611T 786-618T
Micro Dialysis System, Tube-O-Dialyzer™
Tubes Dialysis Tubes
'NO-LOSS' dialyser.
The sample container itself serves as the Tube-O-DIALYZER™, whereby the cap is fitted with a dialysis membrane made from a neutral non binding membrane. Samples are recovered through brief centrifugation. For storing the sample, the dialysis cap can be replaced with a normal cap.
Developed for the dialysis of small but valuable samples. The complete procedure is carried out in special tubes avoiding the need to transfer samples and therefore also avoiding the loss of samples.