VWR®, Centrifugal Filters

Supplier: VWR Collection
516-0227EA 369 EUR
516-0227 516-0228 516-0229 516-0230 516-0231 516-0232 516-0233 516-0234 516-0235 516-0236
VWR®, Centrifugal Filters
Filters Centrifugal Filters
PP insert and receiver tube. MWCO filters are ideal to concentrate, purify and desalt peptides, proteins, oligonucleotides, DNA and RNA.

  • Low binding, typical recovery greater than 90%
  • Sealed membrane
  • Fit rotors that accept 1,5 ml tubes

These 0,2 and 0,45 µm centrifugal filters are used for particulate removal, sample clarification or pre-filtration clean-up of large agarose or acrylamide gel debris prior to further purification with a MWCO device.

Fits rotors that accept 1,5 ml tubes
Order Now

SPECIFICATIONS

Environmentally Preferable
Filtration area 0,28 cm²
Max. RCF 14000 ×g
Dead volume <5 µl
Sample volume Max. 500 µl
Temperature 0...40 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR