VWR® Extraction Thimbles, Grade 501
Supplier: VWR Collection
516-0261EA 117 EUR
516-0261 516-0262 516-0000 516-0252 516-0256 516-0257 516-0258 516-0259 516-0260 516-0254 516-0255 516-0253
VWR® Extraction Thimbles, Grade 501
Extractors Extraction Thimbles
Pure cellulose cotton. Suitable for all types of extractors (Soxhlet, Büchi, Gerhardt and more).
  • Determination of free lipids content in foods and fat content in milk powder
  • Determination of oil content in oleaginous products
  • Extraction and quantification of various components