Multifuge X4F and X4FR Pro Floor Standing Centrifuges and Packages
Supplier: Thermo Scientific
75009530 75009930 75009536 75009936 75017087 75016068
521-3428EA 8813 EUR
521-3428 521-3429 521-3430 521-3431 521-3452 521-3455
Multifuge X4F and X4FR Pro Floor Standing Centrifuges and Packages
Centrifuges Floor-Standing Centrifuges
Ergonomically designed, the Thermo Scientific™ Multifuge X4F/X4FR Pro series floor-standing centrifuges have a touch screen interface that enables easier programming for quicker results and ability to access and track all runs, rotor life and centrifuge health information for maximum versatility and power at every turn.
  • Auto-Lock rotor system
  • ClickSeal™ bucket sealing system
  • SMARTSpin™ technology
  • Space-saving design
  • Energy efficient
  • Capacity up to 4 L, including 196×5/7 ml blood tubes and 40×50 ml conical tubes
Secure locking system allows easy push-More Product Information