Fixed-angle rotor package, TLA-110
Supplier: Beckman Coulter
366735
521-4035EA 15500 EUR
521-4035
Fixed-angle rotor package, TLA-110
Centrifuges Rotors
Major applications: Moderate volume differential sedimentation (pelleting) of subcellular organelles and viruses.