Sorvall™ WX+ Ultracentrifuges
Supplier: SORVALL
75000090 75000100 75000080
521-4305EA 93600 EUR
521-4305 521-4306 521-4304
Sorvall™ WX+ Ultracentrifuges
Centrifuges Floor-Standing Centrifuges
Sorvall™ WX+ Ultracentrifuges, 3 models offering a maximum RCF of up to 802000×g and in a small footprint to maximise space.
  • Available in 80000, 90000 and 100000 min⁻¹ models
  • Open rotor system allows users the flexibility to use other manufacturer's rotors
  • Extensive rotor selection including lightweight, fatigue-resistant Fiberlite™ Carbon fibre rotors
  • Create up to 1000 programmes
Versatile rotor portfolio addresses your high performance processing applications, accommodating tubes from 0,2 to 250 ml providing application flexibility.