Reagent/centrifuge tube with screw cap
Supplier: Greiner Bio-One
188261_500 210261_450 227270_300 210270_300 188271_1000 227261_500 227285_500
391-3477EA 240 EUR
391-3477 391-3460 525-0197 525-0396 391-3450 525-0384 525-0399
Reagent/centrifuge tube with screw cap
Tubes Centrifuge Tubes
PP with HDPE screw cap.
  • Excellent thermal, mechanical and chemical stability
  • Blue graduation and white labelling area
  • DNase-free, RNase-free, human DNA-free, pyrogen-free, non-cytotoxic
Universal tubes for a wide range of applications. With or without skirt. Sterile. Autoclavable without a cap.
Balení: Available in bulk in bags, or packed in boxes.
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001 (241002); ISO 50001 (241002)