VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
525-0629EA 74.5 EUR
525-0629 525-0630 525-0631 525-0632
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Tubes Centrifuge Tubes
PP, high grade gamma resistance, conical, with HDPE flat cap.
  • Easy-to-read black graduations are accurate to ±2%, with 0,5 ml increments (15 ml) or 2,5 ml increments (50 ml)
  • With a large unerasable frosted white printed writing area
  • Both the graduations and writing areas are chloroform-resistant
  • Engraved graduation at the conical bottom of each tube
  • Autoclavable at +121 °C and freezable to −80 °C
  • Leakproof
  • Sustainable design based on virgin resins
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Environmental certification ISO 14001/Certificate No.: 04923E00243R0M