VWR®, Centrifuge Tubes, Borosilicate Glass 3.3
Supplier: VWR Collection
525-1578EA 43 EUR
525-1578 525-1579 525-1580 525-1581 525-1582 525-1583 525-1584 525-1585
VWR®, Centrifuge Tubes, Borosilicate Glass 3.3
Tubes Centrifuge Tubes
Reusable centrifuge tubes made from borosilicate 3.3 glass, available with round or conical bottom, graduated or non graduated.
  • Produced from borosilicate 3.3 glass tubing
  • Highly resistant against chemicals and rapid temperature changes
  • Autoclavable
  • Withstand forces up to 3500 ×g
Graduations and inscriptions in high contrasting blue enamel. With straight rim.
Certifikace: DIN 58970 (only round bottom tubes).