Evaporation system, RapidVap® N₂
Supplier: LABCONCO
7910001 7910011
531-0173EA 26410 EUR
531-0173 531-1077
Evaporation system, RapidVap® N₂
Evaporators Centrifugal Evaporators
An efficient, automatic evaporator system which can be used with a broad range of aggressive chemicals. In RapidVap N₂ systems, a stream of nitrogen or dry gas is directed downward onto the surface of the sample. Nitrogen blow-down reduces the partial pressure directly over the liquid to speed evaporation and help remove the solvent as it evaporates. Nitrogen may be programmed for 2, 4, 6 or all 8 tube locations. Vortex motion and dry heat may also be applied to further accelerate processing.
  • Fast evaporation rates; water evaporates at a rate up to 1,1 ml/minute/tube