'Plunger in needle' syringes, 0,5 to 5 µl
Supplier: Trajan Scientific and Medical
000300 000301 000303 000310 000311 000350 000353 000355 000500 000501 000505 000506 000510 000550 000553 000570 000800 000801 000802 000803 000804 000850 000852
549-0253EA 132 EUR
549-0253 549-0414 549-0415 549-0254 549-0416 549-0255 549-0424 549-0425 549-0258 549-0417 549-0259 549-0418 549-0419 549-0260 549-0261 549-0262 549-0263 549-0420 549-0264 549-0421 549-0422 549-0265 549-0426
'Plunger in needle' syringes, 0,5 to 5 µl
Syringes Syringes for Chromatography
Guaranteed liquid and gas tight to 3,5 bar. The plunger extends into the needle tip. Ideal for dispensing very small liquid volumes.
  • Interchangeable needle and plunger
Replacement plunger/needle kits on request.
* Mentioned Ø and length are those of the needle.