Tightening tools for Very High Pressure (VHP) fittings, Upchurch Scientific®
Supplier: UPCHURCH SCIENTIFIC
VHP-2000 VHP-1000
554-2198EA 1520 EUR
554-2198 554-1999
Tightening tools for Very High Pressure (VHP) fittings, Upchurch Scientific®
Wrenches
  • Configured for optimum torque to ensure a strong connection
  • Prolongs the lifetime of reusable fittings by not over-tightening
  • Available for multiple fitting head styles
Certifikace: Calibrated according to ISO 6789:2003