Avantor® Partisil Avantor® Partisphere, HPLC Columns
Supplier: Avantor
554-5631EA 1150 EUR
554-5631 554-5620 554-5621 554-5622 554-5623 554-5624 554-5625 554-5626 554-5627 554-5628 554-5629 554-5630 554-5632 554-5633 554-5634 554-5635 554-5638 554-5644 554-5645 554-5646 554-5649 554-5651 554-5652 554-5653 554-5654 554-5657 554-5658 554-5659 554-5660 554-5672 554-5674 554-5676 554-5677 554-5678 4227-001M
Avantor® Partisil Avantor® Partisphere, HPLC Columns
Chromatography Columns
Commercially available spherical silicas and it continues to provide and reproducible, high efficiency separations, Avantor® Partisphere® columns are available in a wide range of surface chemistries including two application specific phases.
  • Easy to use
  • Spherical 5 µm porous silica
  • Increased column lifetime