GC capillary column tools
Supplier: Trajan Scientific and Medical
072650 0625011
558-0190EA 51.7 EUR
558-0190 568-0470
GC capillary column tools
Cutters
The capillary cutting tool is used for burr free cutting of fused silica tubing.
  • Reduces likelihood of blockages
  • Ensures square ends for 'Press fit' style connectors
  • Clean entry for on-column injection