Detergents, 'Teepol' L, for glassware
Supplier: TEEPOL
560116C
560116CEA 202 EUR
560116C
Detergents, 'Teepol' L, for glassware
Cleaning Agents Detergents and Surface Cleansers
A neutral liquid detergent particularly good for cleaning laboratory glassware; residual surface activity may be removed by subsequent acid hydrolysis. Avoid contact with eyes, skin and clothes. Working procedure should be designed to minimise exposure to Teepol® L. Dilution rate for cleaning, 15 to 30 ml per 5 litres of water, for general wetting use at a strength of 0,1 to 0,2%.
  • Leaves surfaces clean and bright in all types of water
  • Highly effective wetting agent