Sodium hydroxide 5 N aqueous solution, BAKER ANALYZED® Volumetric, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Caustic soda

5671-02EA 92.22 EUR
5671-02 5671-03
Sodium hydroxide 5 N aqueous solution, BAKER ANALYZED® Volumetric, J.T.Baker®
Sodium hydroxide

J.T.Baker® volumetric and analytical solutions. These solutions are suitable for use in ACS, USP and NF compendial methods and general laboratory applications.


J.T.Baker® volumetric and analytical solutions. These solutions are suitable for use in ACS, USP and NF compendial methods and general laboratory applications.

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: ≤1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Protect from air to avoid absorption of carbon dioxide.
Appearance (Clear, colorless liquid) Passes Test
Normality 4.95 - 5.05
Chloride (Cl) ≤5 ppm
ACS - Heavy Metals (as Pb) ≤1 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Standardization at 25°C traceable to NIST Standard Reference Material.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR