Collecting trays, b.safe
Supplier: Bohlender
R540-01 R540-02 R548-05 R548-10
590-0329EA 52.5 EUR
590-0329 590-0330 590-0331 590-0332
Collecting trays, b.safe
Trays/Dishes/Pots Spillage Trays/Pans
Practical collecting tray made of PE or static dissipative PE-EX. Place your waste canister in a collecting tray. Escaping liquids are collected in case of container exchange or leakages.
  • High-quality HDPE, HDPE-EX
  • Increase your security
  • Liquids are not lost
Collecting trays help to put aside e. g. waste caps or funnels during container exchange without contaminating floor or working area through dripping liquids. In a worst-case scenario they collect escaping liquids from a damaged or spilling waste container and thus increase your own safety.