Laboratory bottles, narrow or wide neck, borosilicate glass, clear or amber, with screw cap
Supplier: S.C.A.T.
501110 501112 501113 501115 501116 501117 501118 501119 501120 501121 501125 107509 101994 101995 101996 101997 101998 501156 501152 501151 501150 501127 501126 199011 501130 501131 501154 501170 501171 501172 501173 501174
590-0832EA 36.9 EUR
590-0832 590-0834 590-0835 590-0836 590-0837 590-0838 590-0839 590-0840 590-0841 590-0842 590-0844 590-0552 590-0500 590-0501 590-0502 590-0503 590-0504 SCTV501156 SCTV501152 SCTV501151 SCTV501150 SCTV501127 SCTV501126 SCTV199011 SCTV501130 SCTV501131 SCTV501154 SCTV501170 SCTV501171 SCTV501172 SCTV501173 SCTV501174
Laboratory bottles, narrow or wide neck, borosilicate glass, clear or amber, with screw cap
Bottles Glass Bottles
Bottles, made of DURAN® borosilicate glass, with blue PP screw cap. Ideal for mixing and storage of culture medium.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Bottles SCTV501130 and SCTV501131 feature a pour ring to facilitate use.
Informace o príslušenství: Bottle tag multipack consists of (2) blue, (2) purple, (2) green, (2) red, (2) orange, (2) yellow, (2) black, and (2) white for a total of 16 pieces.