Hydrochloric acid 32%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

6082.1000EA 36.86 EUR
6082.1000 6082.2500
Hydrochloric acid 32%, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Hydrochloric acid
Formula: HCl
MW: 36.46 g/mol
Melting Pt: –30 °C
Density: 1.18 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Order Now

Specification Test Results

Assay min. 32 %
Ammonium (NH4) max. 1 ppm
Free Chlorine max. 0.5 ppm
Residue after Ignition max. 5 ppm
Sulfate (SO4) max. 1 ppm
Sulfite (SO3) max. 2 ppm
Trace Impurities (in ppm)
Silver (Ag) max. 0.1
Aluminium (Al) max. 0.5
Arsenic (As) max. 0.1
Barium (Ba) max. 0.1
Beryllium (Be) max. 0.1
Calcium (Ca) max. 0.5
Cadmium (Cd) max. 0.1
Cobalt (Co) max. 0.1
Chromium (Cr) max. 0.1
Copper (Cu) max. 0.1
Iron (Fe) max. 0.5
Germanium (Ge) max. 0.1
Potassium (K) max. 0.1
Lithium (Li) max. 0.1
Magnesium (Mg) max. 0.5
Manganese (Mn) max. 0.1
Molybdenum (Mo) max. 0.1
Sodium (Na) max. 0.5
Nickel (Ni) max. 0.1
Lead (Pb) max. 0.1
Strontium (Sr) max. 0.1
Titanium (Ti) max. 0.1
Thallium (Tl) max. 0.1
Vanadium (V) max. 0.1
Zinc (Zn) max. 0.5
Zirconium (Zr) max. 0.1

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR