Precision balances, NewClassic, ML series
Supplier: Mettler - Toledo
11124860 11124890 11124893 11124863 11124865 11124872 11124874 11124875 11124877 11124884 11124886 11124887 11124889 11124892 11124838 11124895 11124902 11124904 11124914 11124916 11124926 11124928 11124929 11124931 11124772 11124862 11124778 11124784 11124809 11124811 11124815 11124817 11124818 11124820 11124824 11124826 11124836
611-2531EA 0 EUR
611-2531 611-2546 METB11124893 METB11124863 METB11124865 611-2532 611-2445 611-2446 METB11124877 611-2447 METB11124886 611-2545 611-2448 611-2449 METB11124838 METB11124895 611-2533 611-2450 611-2451 METB11124916 611-2534 611-2452 611-2535 611-2453 METB11124772 611-2444 METB11124778 METB11124784 611-2439 METB11124811 611-2440 METB11124817 611-2441 METB11124820 611-2442 METB11124826 611-2443
Precision balances, NewClassic, ML series
Balances and Scales
A cost-effective mobile precision balance for accurate and fast weighing tasks.
  • MonoBloc - proven technology for fast and accurate result (except ML802E, ML1502E and ML6001E which have strain gauge technology)
  • Internal Adjustment - a simple keystroke adjustment with an internal weight (except models denoted with suffix E which require external calibration)
  • SmartTrac; graphical display of how much of the range has been used
  • Battery operated or mains (AC) operation for flexibility
  • Backlit display
Informace pro objednávání: OIML certified models and aMore Product Information