Serological pipettes
Supplier: Greiner Bio-One
604181_1000 606180_200 607180_200 710180_1000 760180_200 607107_500 710107_1000 604107_1000 768180_100 606107_500 768160_100 607160_200 760160_200 606160_200 604160_1000 710160_1000 760107_200 760160-TRI_100
612-1011EA 386 EUR
612-1011 612-1013 612-1015 612-1012 612-1017 612-1020 612-9107 612-0677 612-1018 612-1019 612-3328 612-3325 612-3327 612-3324 612-3272 612-3323 GREI760107_200 GREI760160-TRI_100
Serological pipettes
Pipettes Serological Pipettes
High-quality transparent PS, graduated, sterile.
  • Clear and easy-to-read graduation, with a vertical coloured stripe on the 1, 2, 5, 10 and 25-ml pipettes
  • Increased capacity thanks to negative graduation
  • Cotton insert to stop liquids from rising up into the pipetting device
  • Colour code conforms to international standards to facilitate pipette selection
  • Free from DNase, RNase and human DNA, non pyrogenic and non cytotoxic
Balení: Bulk packed in plastic resealable zip sachets, individually wrapped in peelable plastic/paper, or individually packaged inMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Product Transparency and Disclosure Product Transparency and Disclosure Product Transparency and Disclosure Product Transparency and Disclosure
Environmental certification ACT Label (EU 43.9/UK 44.0) ACT Label (EU 39.1/UK 39.2/US 47.0) ACT Label (EU 38.9/UK 39.0/US 46.8) ACT Label (EU 38.6/UK 38.7/US 46.4)