VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
612-1681EA 29 EUR
612-1681 612-1747 612-1682 612-1683 612-1684 612-1685 612-1686 612-1687 612-1688 612-1756 612-1689 612-1690 612-2841 612-2842 612-2843 612-2844 612-2845 612-2846 612-2847 612-2848 612-2849 612-2850 612-2851 621-0058
VWR®, Transfer Pipettes, High Performance
Pipettes Pasteur/Transfer Pipettes
LDPE, disposable. These unbreakable 'all in one' pipettes eliminate the potential hazard of broken glass and exposure to infectious materials. The low affinity surface reduces the loss of cells and valuable proteins due to binding. They work well whenever there is a need for quick, safe transfer of fluids.
  • Non toxic and inert to biological fluids and most acids
  • Can be heat-sealed and frozen at very low temperatures such as in liquid nitrogen
  • Available sterile or non sterile
  • Sterility assurance level is SAL 10⁻⁶
Balení: Sterile pipettes are packed inMore Product Information