Pipette controller, micro-classic
Supplier: Brand
25900
612-1912EA 55.4 EUR
612-1912
Pipette controller, micro-classic
Pipettes Pipette Controllers
Suitable for disposable micropipettes with ring mark and other small volume pipettes up to 1 ml (e.g. blood diluting pipettes) with aspiration end diameter maximum 5 mm.
  • Ergonomic shape and simple handling offering comfort and convenience for this job
  • For IVF and medical laboratories
  • Minimises the risk of contamination when working with infectious material
  • Adapter for right- and left-handed operation and suction tube are autoclavable