Burettes, straight stopcock
Supplier: HIRSCHMANN
3120160 3120170 3120175
612-3541EA 106 EUR
612-3541 612-3542 612-3543
Burettes, straight stopcock
Burettes
DURAN®, brown glass, class B.
  • Calibrated to deliver 'Ex'
  • Short marks at major graduations, white graduations
  • Glass key
Scale length : 500 mm
Certifikace: DIN EN ISO 385