Volumetric flasks, class B, red markings
Supplier: VITLAB
67695
612-3916EA 24.9 EUR
612-3916
Volumetric flasks, class B, red markings
Flasks Volumetric Flasks
PMP, translucent, with PP NS-stopper.
  • Tolerances are approximately 20% better than class B according to DIN EN ISO 1042
  • Individually ring marked
  • With NS stoppers
Certifikace: DIN EN ISO 1042